Thermo Scientific Chemicals

Pentaerythritol, 98%, Thermo Scientific Chemicals

Catalog number: 129870050
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Pentaerythritol, 98%, Thermo Scientific Chemicals

Catalog number: 129870050
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 129870050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
115-77-5
IUPAC Name
2,2-bis(hydroxymethyl)propane-1,3-diol
Molecular Formula
C5H12O4
InChI Key
WXZMFSXDPGVJKK-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)CO
Appearance (Color)
White
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Crystalline powder or crystals
Melting point
257°C to 265°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text