Pentafluorothiophenol, 97%, Thermo Scientific Chemicals
Pentafluorothiophenol, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Pentafluorothiophenol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 mL
5 mL
Packaging:
Glass bottle
Catalog number 129920250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepentafluorobenzene-1-thiol
Molecular FormulaC6HF5S
InChI KeyUVAMFBJPMUMURT-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(S)C(F)=C1F
Molecular Weight (g/mol)200.13
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4630 to 1.4650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only