Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:For superior PCR performance, upgrade to Invitrogen's new Platinum II Taq Hot-start DNA Polymerase.
Invitrogen™

Platinum™ Green Hot Start PCR Master Mix (2X)

Catalog number: 13001013
Have Questions?
Invitrogen™

Platinum™ Green Hot Start PCR Master Mix (2X)

Catalog number: 13001013
Have Questions?
Catalog Number
13001013
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
13001013200 reactions
-
1300101250 reactions
-
130010141000 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Invitrogen Platinum Green Hot Start PCR Master Mix offers Platinum Taq DNA polymerase in an optimized PCR buffer with magnesium and dNTPs for convenient PCR setup. It retains all the features of Platinum Taq DNA Polymerase: specificity, robustness, and reliability. Platinum Green Hot Start PCR Master Mix is supplemented with two tracking dyes for the convenience of direct gel loading of PCR products.

Product features:
• High specificity and increased yields with antibody-mediated hot-start PCR
• Convenient room-temperature reaction setup
• Direct gel loading with green PCR master mix
• Versatile formulation for broad range of amplicons

About Platinum Taq DNA Polymerase
Platinum Taq DNA Polymerase is a recombinant Taq DNA polymerase complexed with proprietary antibodies that inhibit enzyme activity at room temperature, reducing nonspecific target amplification at lower temperatures. The antibodies dissociate during the initial PCR denaturation step and DNA polymerase regains its full activity. Just as with Taq DNA Polymerase, Platinum Taq DNA Polymerase has a non-template-dependent terminal transferase activity that adds a 3' deoxyadenosine to product ends and has a 5'→3' exonuclease activity.

Using Platinum Green Hot Start PCR Master Mix
Convenient Platinum Green Hot Start PCR Master Mix provides a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. The two tracking dyes in Platinum Green Hot Start PCR Master Mix enable direct gel loading of PCR products, thus eliminating tedious steps of dye addition. Platinum Green Hot Start PCR Master Mix is supplied with a separate vial of Platinum GC Enhancer formulated for specific amplification and improved yields of GC-rich targets. Use Platinum Green Hot Start PCR Master Mix in PCR applications such as genotyping, gene expression profiling, and next-generation sequencing.

Note: For superior PCR performance, we recommend next generation enzyme Platinum II Taq Hot-start DNA Polymerase. Platinum II Taq Hot-Start DNA Polymerase is designed for universal primer annealing and fast, easy PCR with its unique combination of innovative buffer, high-performance engineered Taq DNA polymerase, and superior hot-start technology. Platinum II Hot-Start Green PCR Master Mix contains Platinum II Taq Hot-Start DNA Polymerase in a ready-to-use mixture with Platinum II PCR buffer and dNTPs, thus reducing the number of pipetting steps during PCR reaction setup. It additionally contains a density reagent and two tracking dyes for direct loading of PCR products on gels.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Tube
GC-Rich PCR Performance
Medium
Polymerase
Platinum Taq DNA Polymerase
Reaction Speed
Standard
Label or Dye
Green
Product Line
Platinum™
Product Type
Hot Start PCR Master Mix
Shipping Condition
Dry Ice
Sufficient For
200 Reactions
Concentration
2X
Fidelity (vs. Taq)
1 X
Hot Start
Built-In Hot Start
No. of Reactions
200 Reactions
Overhang
3'-A
Reaction Format
SuperMix or Master Mix
Size (Final Product)
5 kb or less
Starting Material
DNA

Contents & Storage

Each box contains:
• 4 x 1.25 mL Platinum Green Hot Start PCR 2X Master Mix
• 2 x 1.25 mL Platinum GC Enhancer
• 4 x 1.25 mL water, nuclease-free

Store at -20°C in a non-frost-free freezer.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text