Perchloromethyl mercaptan, 97%, Thermo Scientific Chemicals
Perchloromethyl mercaptan, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perchloromethyl mercaptan, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 130030250
Price (USD)/ Each
108.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
108.00
Add to cart
Perchloromethyl mercaptan, 97%, Thermo Scientific Chemicals
Catalog number130030250
Price (USD)/ Each
108.00
-
Add to cart
Chemical Identifiers
CAS594-42-3
IUPAC Nametrichloro(chlorosulfanyl)methane
Molecular FormulaCCl4S
InChI KeyRYFZYYUIAZYQLC-UHFFFAOYSA-N
SMILESClSC(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear yellow
Refractive index1.5370 to 1.5410 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only