Perylene, 99+%, Thermo Scientific Chemicals
Perylene, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Perylene, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 130070050
Price (USD)/ Each
116.65
Online exclusive
137.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
116.65
Online exclusive
137.00 
Save 20.35 (15%)
Add to cart
Perylene, 99+%, Thermo Scientific Chemicals
Catalog number130070050
Price (USD)/ Each
116.65
Online exclusive
137.00 
Save 20.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameperylene
Molecular FormulaC20H12
InChI KeyCSHWQDPOILHKBI-UHFFFAOYSA-N
SMILESC1=CC2=C3C(C=CC=C3C3=C4C(C=CC=C24)=CC=C3)=C1
Molecular Weight (g/mol)252.32
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Ochre to brown
Melting point277°C to 280°C
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only