Chemical Identifiers
CAS128-69-8
IUPAC Name7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
Molecular FormulaC24H8O6
InChI KeyCLYVDMAATCIVBF-UHFFFAOYSA-N
SMILESO=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56
View more