1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130130250
Price (USD)/ Each
151.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
178.00 
Save 26.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
151.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
178.00 
Save 26.35 (15%)
Add to cart
1,10-Phenanthroline monohydrate, 99+%, pure, Thermo Scientific Chemicals
Catalog number130130250
Price (USD)/ Each
151.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
178.00 
Save 26.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,10-phenanthroline hydrate
Molecular FormulaC12H10N2O
InChI KeyPPQJCISYYXZCAE-UHFFFAOYSA-N
SMILESO.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
Molecular Weight (g/mol)198.23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder or needles
Infrared spectrumConforms
Titration with HClO499.0 to 101.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only