1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, Thermo Scientific Chemicals
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130140250
Price (USD)/ Each
132.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
156.00 
Save 23.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
132.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
156.00 
Save 23.35 (15%)
Add to cart
1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, Thermo Scientific Chemicals
Catalog number130140250
Price (USD)/ Each
132.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
156.00 
Save 23.35 (15%)
-
Add to cart
Chemical Identifiers
CAS18851-33-7
IUPAC Name1,10-phenanthroline hydrate hydrochloride
Molecular FormulaC12H11ClN2O
InChI KeyNDLHUHRGAIHALB-UHFFFAOYSA-N
SMILESO.Cl.C1=CN=C2C(C=CC3=CC=CN=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Sulfated ash=<0.2 %
Appearance (Color)White to light yellow to light purple
Infrared spectrumConforms
Appearance (Form)Powder or crystals
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only