Phenazine, 98%, Thermo Scientific Chemicals
Phenazine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenazine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130150050
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.40 
Save 6.75 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.40 
Save 6.75 (15%)
Add to cart
Phenazine, 98%, Thermo Scientific Chemicals
Catalog number130150050
Price (USD)/ Each
39.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
46.40 
Save 6.75 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namephenazine
Molecular FormulaC12H8N2
InChI KeyPCNDJXKNXGMECE-UHFFFAOYSA-N
SMILESC1=CC2=NC3=CC=CC=C3N=C2C=C1
Molecular Weight (g/mol)180.21
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Yellow to brown
Melting point173°C to 177°C
Appearance (Form)Crystalline powder or crystals or needles
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only