Phenazine methosulfate, 98%, Thermo Scientific Chemicals
Phenazine methosulfate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenazine methosulfate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130160100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS299-11-6
IUPAC Name5-methylphenazin-5-ium methyl sulfate
Molecular FormulaC14H14N2O4S
InChI KeyRXGJTUSBYWCRBK-UHFFFAOYSA-M
SMILESCOS([O-])(=O)=O.C[N+]1=C2C=CC=CC2=NC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to khaki to brown
Appearance (Form)Crystalline powder
UV spectrophotometry>=97.5 % (0.01g/l H2O, 1cm cell, lambda max 387.5 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only