1x1 image pixel for data collection
BioLite™ Cell Culture Treated Dishes
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
BioLite™ Cell Culture Treated Dishes
Thermo Scientific™

BioLite™ Cell Culture Treated Dishes

Perform general cell culture procedures more economically with Thermo Scientific™ BioLite™ Cell Culture Treated Dishes that are guaranteed to be sterile, non-pyrogenic, and non-cytotoxic.
Promo ImagePromotion
Promo code:RCELL51
Stock your lab with plasticware essentials so that you can culture with confidence
From now until Dec 31, get 51% off various cell culture plasticware like dishes, flasks, plates and tubes that help achieve reproducible resultsLearn More
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
130182Promo Image100 mm
130180Promo Image35 mm
130181Promo Image60 mm
130183Promo Image150 mm
Catalog number 130182
Price (USD)
349.65
Online Exclusive
387.00
Save 37.35 (10%)
Case of 200
Add to cart
Diameter (Metric):
100 mm
Price (USD)
349.65
Online Exclusive
387.00
Save 37.35 (10%)
Case of 200
Add to cart
Experience optimal performance with wide selections in size, Thermo Scientific™ BioLite™ cell culture treated dishes suit a multitude of experimental needs while providing cost efficiency for a wide selection of applications in your cell culture routine.
• Cell culture treated
• Sterile per ISO 11137, SAL 10-6
• USP Class VI and non-cytotoxic
• Non-pyrogenic

Specifications
Height (Metric) Interior16 mm
No. per Case200
No. per Pack10
ShapeRound
TypeCell Culture Dish
Volume (Metric) Working12.5 mL
Bottom ShapeFlat
Capacity (Metric)12.5 mL (Well)
Culture Area60.8 sq. cm
Diameter (Metric)100 mm
Height (Metric)20 mm
MaterialPolyethylene
SterilitySterile
Surface TreatmentCell Culture
Unit SizeCase of 200

Frequently asked questions (FAQs)

What is the recommended working volume of Nunc adherent cell culture products?

This depends largely on the cell line and working conditions (considerations include headspace for gas exchange, supplements used, and optimal feeding schedule, among others) but we do offer guidelines for development purposes (0.3 to 0.5 mL per cm2) . Note that these volumes are approximate and may have to be modified for best performance. T25 Flask = 7 mL, T75 Flask = 25 mL, T175 Flask = 55 mL, T225 Flask = 70 mL, 300cm2 Flask = 100 mL, 500 cm2 TripleFlask = 200 mL, 35 mm round culture dish = 3 mL, 60 mm round culture dish = 5 mL, 100 mm round culture dish = 12 mL, 150 mm round culture dish = 35 mL, 4 (rectangular) well Multidish = 5 mL, 8 (rectangular) well Multidish = 3 mL, 4 (round) well Multidish = 1 mL, 6 well Multidish = 3 mL, 12 well Multidish = 2 mL, 24 well Multidish = 1 mL, 48 well Multidish = 0.5 mL.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link