Phenethyl alcohol, 99%, Thermo Scientific Chemicals
Phenethyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenethyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
5 mL
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 130182500
Price (USD)/ Each
61.65
Online exclusive
68.30 
Save 6.65 (10%)
-
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
61.65
Online exclusive
68.30 
Save 6.65 (10%)
Add to cart
Phenethyl alcohol, 99%, Thermo Scientific Chemicals
Catalog number130182500
Price (USD)/ Each
61.65
Online exclusive
68.30 
Save 6.65 (10%)
-
Add to cart
Chemical Identifiers
CAS60-12-8
IUPAC Name2-phenylethan-1-ol
Molecular FormulaC8H10O
InChI KeyWRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.5300 to 1.5350 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only