Phenothiazine, 99%
Phenothiazine, 99%
Phenothiazine, 99%
Thermo Scientific Chemicals

Phenothiazine, 99%

CAS: 92-84-2 | C12H9NS | 199.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 130210010
Price (USD)
101.00
Each
In stock
Add to cart
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
101.00
Each
Add to cart
Chemical Identifiers
CAS92-84-2
IUPAC Name10H-phenothiazine
Molecular FormulaC12H9NS
InChI KeyWJFKNYWRSNBZNX-UHFFFAOYSA-N
SMILESN1C2=CC=CC=C2SC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Beads or prills
HPLC>=98.5 %
Appearance (Color)Yellow
Melting point>=184°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471147Certificate of AnalysisMay 17, 2025130210050, 130215000, 130210010
A0471148Certificate of AnalysisMay 17, 2025130210050, 130215000, 130210010
A0472374Certificate of AnalysisMay 17, 2025130210050, 130215000, 130210010
A0466796Certificate of AnalysisOct 15, 2024130210050, 130215000, 130210010
A0463349Certificate of AnalysisMay 30, 2024130210050, 130215000, 130210010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection