Thermo Scientific Chemicals

Phenothiazine, 99%, Thermo Scientific Chemicals

Catalog number: 130215000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Phenothiazine, 99%, Thermo Scientific Chemicals

Catalog number: 130215000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 130215000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
92-84-2
IUPAC Name
10H-phenothiazine
Molecular Formula
C12H9NS
InChI Key
WJFKNYWRSNBZNX-UHFFFAOYSA-N
SMILES
N1C2=CC=CC=C2SC2=CC=CC=C12
Appearance (Color)
Yellow
Melting point
>=184°C
Appearance (Form)
Beads or prills
HPLC
>=98.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text