Phenoxazine, 97%, Thermo Scientific Chemicals
Phenoxazine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenoxazine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 130220250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS135-67-1
IUPAC Name10H-phenoxazine
Molecular FormulaC12H9NO
InChI KeyTZMSYXZUNZXBOL-UHFFFAOYSA-N
SMILESN1C2=CC=CC=C2OC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point154°C to 159°C
Appearance (Color)Beige to grey
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only