2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%
2-Phenoxyethanol, 99%
Thermo Scientific Chemicals

2-Phenoxyethanol, 99%

2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C | CAS: 122-99-6 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
250 mL
Packaging:
View all
Glass bottle
Catalog number 130240010
Price (USD)
52.65
Special offer
Online exclusive
Ends: 26-Sep-2025
64.50
Save 11.85 (18%)
Each
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 26-Sep-2025
64.50
Save 11.85 (18%)
Each
Add to cart
Chemical Identifiers
CAS122-99-6
IUPAC Name2-phenoxyethan-1-ol
Molecular FormulaC8H10O2
InChI KeyQCDWFXQBSFUVSP-UHFFFAOYSA-N
SMILESOCCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.5360 to 1.5400 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link