Thermo Scientific Chemicals

L-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 130320500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

L-Phenylalanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 130320500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130320500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7524-50-7
IUPAC Name
hydrogen methyl 2-amino-3-phenylpropanoate chloride
Molecular Formula
C10H14ClNO2
InChI Key
SWVMLNPDTIFDDY-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1
Appearance (Color)
White to off-white
Titration Argentometric
>=97.5 %
Appearance (Form)
Crystalline powder
Specific optical rotation
+37° to +39° (20°C, 589 nm) (c=2, C2H5OH)
Melting point
156°C to 160°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text