Phenylboronic acid, 98+%, may contain varying amounts of anhydride
Phenylboronic acid, 98+%, may contain varying amounts of anhydride
Phenylboronic acid, 98+%, may contain varying amounts of anhydride
Phenylboronic acid, 98+%, may contain varying amounts of anhydride
Thermo Scientific Chemicals

Phenylboronic acid, 98+%, may contain varying amounts of anhydride

CAS: 98-80-6 | C6H7BO2 | 121.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
10 g
Packaging:
Glass bottle
Catalog number 130360500
Price (EUR)
138,40
Online Exclusive
173,00
Save 34,60 (20%)
Each
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
138,40
Online Exclusive
173,00
Save 34,60 (20%)
Each
Add to cart
Chemical Identifiers
CAS98-80-6
IUPAC Namephenylboronic acid
Molecular FormulaC6H7BO2
InChI KeyHXITXNWTGFUOAU-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or powder or crystals
HPLC>=98.0 %
Appearance (Color)White to almost white
Infrared spectrumConforms
Melting point214°C to 220°C
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0450475Certificate of AnalysisMay 17, 2025130360500, 130362500, 130360100
A0471800Certificate of AnalysisMay 17, 2025130360500, 130362500, 130360100
A0423703Certificate of AnalysisOct 07, 2024130360500, 130362500, 130360100
A0463890Certificate of AnalysisJun 24, 2024130360500, 130362500, 130360100
A0457674Certificate of AnalysisNov 20, 2023130360500, 130362500, 130360100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Label Elements
Signal Word

Warning

Hazard Statements

H302 - Harmful if swallowed

Precautionary Statements

P264 - Wash face, hands and any exposed skin thoroughly after handling

P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

P312 - Call a POISON CENTER or doctor if you feel unwell

Share catalog number, name or link

1x1 image pixel for data collection