Thermo Scientific Chemicals

4-Phenylbutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 130380250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Phenylbutyric acid, 99%, Thermo Scientific Chemicals

Catalog number: 130380250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130380250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1821-12-1
IUPAC Name
4-phenylbutanoic acid
Molecular Formula
C10H12O2
InChI Key
OBKXEAXTFZPCHS-UHFFFAOYSA-N
SMILES
OC(=O)CCCC1=CC=CC=C1
Appearance (Form)
Powder or crystals and/or chunks or fused solid
Titration with NaOH
98.5 to 101.5 %
Appearance (Color)
White to light yellow to beige
Melting point
49°C to 53°C
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text