o-Phenylenediacetonitrile, 99+%, Thermo Scientific Chemicals
o-Phenylenediacetonitrile, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Phenylenediacetonitrile, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 130520500
Price (USD)/ Each
136.00
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
136.00
Add to cart
o-Phenylenediacetonitrile, 99+%, Thermo Scientific Chemicals
Catalog number130520500
Price (USD)/ Each
136.00
-
Add to cart
Chemical Identifiers
CAS613-73-0
IUPAC Name2-[2-(cyanomethyl)phenyl]acetonitrile
Molecular FormulaC10H8N2
InChI KeyFWPFXBANOKKNBR-UHFFFAOYSA-N
SMILESN#CCC1=CC=CC=C1CC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
AppearanceMay contain yellow to orange chunks
Infrared spectrumConforms
Appearance (Color)White to yellow to beige
Melting point57°C to 61°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only