m-Phenylenediamine, 99+%, Thermo Scientific Chemicals
m-Phenylenediamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

m-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 130565000
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.10 
Save 6.45 (14%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.10 
Save 6.45 (14%)
Add to cart
m-Phenylenediamine, 99+%, Thermo Scientific Chemicals
Catalog number130565000
Price (USD)/ Each
40.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
47.10 
Save 6.45 (14%)
-
Add to cart
Chemical Identifiers
IUPAC Namebenzene-1,3-diamine
Molecular FormulaC6H8N2
InChI KeyWZCQRUWWHSTZEM-UHFFFAOYSA-N
SMILESNC1=CC(N)=CC=C1
Molecular Weight (g/mol)108.14
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Melting point62°C to 65°C
Appearance (Color)White to tan to grey
Appearance (Form)Flakes or pellets
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only