Thermo Scientific Chemicals

p-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Catalog number: 130570025
2.5 kg, Plastic drum, Each
Thermo Scientific Chemicals

p-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Catalog number: 130570025
2.5 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 130570025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
106-50-3
IUPAC Name
benzene-1,4-diamine
Molecular Formula
C6H8N2
InChI Key
CBCKQZAAMUWICA-UHFFFAOYSA-N
SMILES
NC1=CC=C(N)C=C1
Appearance (Color)
White to grey to brown to purple
GC
>=99.0 %
Appearance (Form)
Powder or flakes
Infrared spectrum
Conforms
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text