1-Phenyl-1,2-ethanediol, 97%
1-Phenyl-1,2-ethanediol, 97%
1-Phenyl-1,2-ethanediol, 97%
1-Phenyl-1,2-ethanediol, 97%
Thermo Scientific Chemicals

1-Phenyl-1,2-ethanediol, 97%

CAS: 93-56-1 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 130591000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS93-56-1
IUPAC Name1-phenylethane-1,2-diol
Molecular FormulaC8H10O2
InChI KeyPWMWNFMRSKOCEY-UHFFFAOYNA-N
SMILESOCC(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light beige
Appearance (Form)Powder or crystalline powder or crystals
Infrared spectrumConforms
Melting point62°C to 68°C
GC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0461022Certificate of AnalysisMar 19, 2024130590250, 130590050, 130591000
A0410954Certificate of AnalysisFeb 04, 2022130590250, 130590050, 130591000
A0401792Certificate of AnalysisApr 28, 2021130590250, 130590050, 130591000
3 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection