Thermo Scientific Chemicals

N-Phenylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: 130610100
10 mL, Glass bottle, Each
Thermo Scientific Chemicals

N-Phenylethylenediamine, 99%, Thermo Scientific Chemicals

Catalog number: 130610100
10 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 130610100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1664-40-0
IUPAC Name
N1-phenylethane-1,2-diamine
Molecular Formula
C8H12N2
InChI Key
OCIDXARMXNJACB-UHFFFAOYSA-N
SMILES
NCCNC1=CC=CC=C1
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Titration with HClO4
>=98.5 %
Refractive index
1.5855 to 1.5875 (20°C, 589 nm)
Appearance (Color)
Clear yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text