Thermo Scientific Chemicals

N-Phenylmaleimide, 97%, Thermo Scientific Chemicals

Catalog number: 130710250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

N-Phenylmaleimide, 97%, Thermo Scientific Chemicals

Catalog number: 130710250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130710250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
941-69-5
IUPAC Name
1-phenyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular Formula
C10H7NO2
InChI Key
HIDBROSJWZYGSZ-UHFFFAOYSA-N
SMILES
O=C1C=CC(=O)N1C1=CC=CC=C1
Appearance (Form)
Crystalline powder or flakes
Melting point
86°C to 91°C
Infrared spectrum
Conforms
GC
>=96.0 %
Appearance (Color)
Yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text