Phenylphosphonic acid, 98%, Thermo Scientific Chemicals
Phenylphosphonic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenylphosphonic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 130791000
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
Add to cart
Phenylphosphonic acid, 98%, Thermo Scientific Chemicals
Catalog number130791000
Price (USD)/ Each
47.65
Online exclusive
53.20 
Save 5.55 (10%)
-
Add to cart
Chemical Identifiers
CAS1571-33-1
IUPAC Namephenylphosphonic acid
Molecular FormulaC6H7O3P
InChI KeyQLZHNIAADXEJJP-UHFFFAOYSA-N
SMILESOP(O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point158°C to 165°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only