Thermo Scientific Chemicals

Phenylphosphonic dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 130800010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Phenylphosphonic dichloride, 97%, Thermo Scientific Chemicals

Catalog number: 130800010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 130800010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
824-72-6
IUPAC Name
phenylphosphonoyl dichloride
Molecular Formula
C6H5Cl2OP
InChI Key
IBDMRHDXAQZJAP-UHFFFAOYSA-N
SMILES
ClP(Cl)(=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow-brown
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.5570 to 1.5610 (20°C, 589 nm)
Titration Argentometric
96.0 to 104.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text