Thermo Scientific Chemicals

2-Phenyl-2-propanol, 99%, Thermo Scientific Chemicals

Catalog number: 130831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Phenyl-2-propanol, 99%, Thermo Scientific Chemicals

Catalog number: 130831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130831000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
617-94-7
IUPAC Name
2-phenylpropan-2-ol
Molecular Formula
C9H12O
InChI Key
BDCFWIDZNLCTMF-UHFFFAOYSA-N
SMILES
CC(C)(O)C1=CC=CC=C1
Appearance (Color)
Colorless to white to light yellow
Appearance (Form)
or liquid as melt
Melting point
29°C to 35°C
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text