Thermo Scientific Chemicals

DL-2-Phenylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 130870250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-2-Phenylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: 130870250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130870250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
492-37-5
IUPAC Name
2-phenylpropanoic acid
Molecular Formula
C9H10O2
InChI Key
YPGCWEMNNLXISK-UHFFFAOYNA-N
SMILES
CC(C(O)=O)C1=CC=CC=C1
Infrared spectrum
Conforms
Titration with NaOH
97.5 to 102.5 %
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text