4-Phenylpyridine, 99%, Thermo Scientific Chemicals
4-Phenylpyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Phenylpyridine, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 130911000
Price (USD)/ Each
835.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
835.00
Add to cart
4-Phenylpyridine, 99%, Thermo Scientific Chemicals
Catalog number130911000
Price (USD)/ Each
835.00
-
Add to cart
Chemical Identifiers
IUPAC Name4-phenylpyridine
Molecular FormulaC11H9N
InChI KeyJVZRCNQLWOELDU-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)C1=CC=NC=C1
Molecular Weight (g/mol)155.20
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point71°C to 75°C
Appearance (Color)White to light yellow to beige
Appearance (Form)Powder or crystals
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only