Thermo Scientific Chemicals

DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 130941000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

DL-Phenylsuccinic acid, 98+%, Thermo Scientific Chemicals

Catalog number: 130941000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 130941000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
635-51-8
IUPAC Name
2-phenylbutanedioic acid
Molecular Formula
C10H10O4
InChI Key
LVFFZQQWIZURIO-UHFFFAOYNA-N
SMILES
OC(=O)CC(C(O)=O)C1=CC=CC=C1
Infrared spectrum
Conforms
Melting point
165°C to 169°C
Appearance (Color)
White to cream
Appearance (Form)
Crystalline powder
Titration with NaOH
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text