Thermo Scientific Chemicals

Phenyl sulfide, 99%, Thermo Scientific Chemicals

Catalog number: 130951000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Phenyl sulfide, 99%, Thermo Scientific Chemicals

Catalog number: 130951000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 130951000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
139-66-2
IUPAC Name
(phenylsulfanyl)benzene
Molecular Formula
C12H10S
InChI Key
LTYMSROWYAPPGB-UHFFFAOYSA-N
SMILES
S(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Form)
Liquid
GC
>=98.5 %
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
Refractive index
1.6317 to 1.6337 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text