Phenyl sulfone, 97%, Thermo Scientific Chemicals
Phenyl sulfone, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl sulfone, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 130962500
Price (USD)/ Each
62.30
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
62.30
Add to cart
Phenyl sulfone, 97%, Thermo Scientific Chemicals
Catalog number130962500
Price (USD)/ Each
62.30
-
Add to cart
Chemical Identifiers
IUPAC Name(benzenesulfonyl)benzene
Molecular FormulaC12H10O2S
InChI KeyKZTYYGOKRVBIMI-UHFFFAOYSA-N
SMILESO=S(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)218.27
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Melting point125°C to 129°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only