Phenyl sulfoxide, 97%, Thermo Scientific Chemicals
Phenyl sulfoxide, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenyl sulfoxide, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 130971000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS945-51-7
IUPAC Name(benzenesulfinyl)benzene
Molecular FormulaC12H10OS
InChI KeyJJHHIJFTHRNPIK-UHFFFAOYSA-N
SMILESO=S(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.5 % (K.F.)
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point66°C to 71°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only