Phenylurea, 97%, Thermo Scientific Chemicals
Phenylurea, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phenylurea, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
Packaging:
Plastic bottle
Catalog number 131021000
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.90 
Save 6.25 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.90 
Save 6.25 (15%)
Add to cart
Phenylurea, 97%, Thermo Scientific Chemicals
Catalog number131021000
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
41.90 
Save 6.25 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namephenylurea
Molecular FormulaC7H8N2O
InChI KeyLUBJCRLGQSPQNN-UHFFFAOYSA-N
SMILESNC(=O)NC1=CC=CC=C1
Molecular Weight (g/mol)136.15
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Melting point143°C to 149°C
Appearance (Color)Almost white to beige
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only