Phthalhydrazide, 99%, Thermo Scientific Chemicals
Phthalhydrazide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalhydrazide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 131061000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1445-69-8
IUPAC Name1,4,4a,8a-tetrahydrophthalazine-1,4-dione
Molecular FormulaC8H6N2O2
InChI KeyJNQUJPBFSQETPT-UHFFFAOYNA-N
SMILESO=C1N=NC(=O)C2C=CC=CC12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or chunks
Loss on drying=<1 %
Appearance (Color)White
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 % (On dried substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only