Phthalimide, 99%, Thermo Scientific Chemicals
Phthalimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phthalimide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 131100010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS85-41-6
IUPAC Name2,3-dihydro-1H-isoindole-1,3-dione
Molecular FormulaC8H5NO2
InChI KeyXKJCHHZQLQNZHY-UHFFFAOYSA-N
SMILESO=C1NC(=O)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point232°C to 236°C
HPLC>=98.5 %
Appearance (Color)White to light yellow
Appearance (Form)Powder or flakes
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only