Thermo Scientific Chemicals

Phthalimide, 99%, Thermo Scientific Chemicals

Catalog number: 131101000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Phthalimide, 99%, Thermo Scientific Chemicals

Catalog number: 131101000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 131101000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-41-6
IUPAC Name
2,3-dihydro-1H-isoindole-1,3-dione
Molecular Formula
C8H5NO2
InChI Key
XKJCHHZQLQNZHY-UHFFFAOYSA-N
SMILES
O=C1NC(=O)C2=CC=CC=C12
Melting point
232°C to 236°C
HPLC
>=98.5 %
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or flakes
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text