o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals
o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals
o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals
o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
1311100505 mLGlass bottle
Catalog number 131110050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS88-95-9
IUPAC Namebenzene-1,2-dicarbonyl dichloride
Molecular FormulaC8H4Cl2O2
InChI KeyFYXKZNLBZKRYSS-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=CC=C1C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5674 to 1.5694 (20°C, 589 nm)
Appearance (Color)Clear yellow
Titration RCOCl>=88 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link