Thermo Scientific Chemicals

Picolinic acid N-oxide, 97%, Thermo Scientific Chemicals

Catalog number: 131180250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Picolinic acid N-oxide, 97%, Thermo Scientific Chemicals

Catalog number: 131180250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131180250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
824-40-8
IUPAC Name
2-carboxypyridin-1-ium-1-olate
Molecular Formula
C6H5NO3
InChI Key
FHYMLBVGNFVFBT-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=CC=[N+]1[O-]
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Water
=<0.5 % (K.F.)
HPLC
>=96.0 %
Appearance (Color)
White to beige

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text