Thermo Scientific Chemicals

4-Picolyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 131200100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Picolyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Catalog number: 131200100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131200100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1822-51-1
IUPAC Name
hydrogen 4-(chloromethyl)pyridine chloride
Molecular Formula
C6H7Cl2N
InChI Key
ZDHKVKPZQKYREU-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClCC1=CC=NC=C1
Appearance (Color)
Yellow to orange
Titration with HClO4
>=96.0 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text