4-Picolyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
4-Picolyl chloride hydrochloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Picolyl chloride hydrochloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
Glass bottle
Catalog number 131201000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namehydrogen 4-(chloromethyl)pyridine chloride
Molecular FormulaC6H7Cl2N
InChI KeyZDHKVKPZQKYREU-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClCC1=CC=NC=C1
Molecular Weight (g/mol)164.03
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Titration with HClO4>=96.0 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only