Thermo Scientific Chemicals

Picrotoxin, 98%, Thermo Scientific Chemicals

Catalog number: 131210010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Picrotoxin, 98%, Thermo Scientific Chemicals

Catalog number: 131210010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131210010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
124-87-8
IUPAC Name
(1R,3R,5S,8S,13R,14S)-1-hydroxy-14-(2-hydroxypropan-2-yl)-13-methyl-4,7,10-trioxapentacyclo[6.4.1.1⁹,¹².0³,⁵.0⁵,¹³]tetradecane-6,11-dione; (1R,5S,8S,13R,14R)-1-hydroxy-13-methyl-14-(prop-1-en-2-yl)-4,7,10-trioxapentacyclo[6.4.1.1⁹,¹².0³,⁵.0⁵,¹³]tetradecane-6,11-dione
Molecular Formula
C30H34O13
InChI Key
VJKUPQSHOVKBCO-ZTYBEOBUSA-N
SMILES
CC(=C)[C@@H]1C2OC(=O)C1[C@]1(O)CC3O[C@]33C(=O)O[C@H]2[C@]13C.CC(C)(O)[C@@H]1C2OC(=O)C1[C@]1(O)C[C@H]3O[C@]33C(=O)O[C@H]2[C@]13C
Appearance (Color)
White to light yellow
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
Melting point
198°C to 204°C
HPLC
>=94 % (Mixture of Picrotoxinin and Picrotin)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text