Thermo Scientific Chemicals

Pimelic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 131230250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Pimelic Acid, 98%, Thermo Scientific Chemicals

Catalog number: 131230250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 131230250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-16-0
IUPAC Name
heptanedioic acid
Molecular Formula
C7H12O4
InChI Key
WLJVNTCWHIRURA-UHFFFAOYSA-N
SMILES
OC(=O)CCCCCC(O)=O
Appearance (Color)
White to light beige
Melting point
103°C to 106°C
Titration with NaOH
97.5 to 102.5 %
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text