(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1R)-(+)-alpha-Pinene, 98%, 80% ee, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 131262500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7785-70-8
IUPAC Name(1R,5R)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular FormulaC10H16
InChI KeyGRWFGVWFFZKLTI-RKDXNWHRSA-N
SMILESCC1=CC[C@@H]2C[C@H]1C2(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Specific gravity(20°C) 0.855 to 0.865
Infrared spectrumConforms
GC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only