Thermo Scientific Chemicals

(1S)-(-)-α-Pinene, 98%, Thermo Scientific Chemicals

Catalog number: 131270025
2.5 L, Glass bottle, Each
Thermo Scientific Chemicals

(1S)-(-)-α-Pinene, 98%, Thermo Scientific Chemicals

Catalog number: 131270025
2.5 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 131270025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7785-26-4
IUPAC Name
(1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-ene
Molecular Formula
C10H16
InChI Key
GRWFGVWFFZKLTI-IUCAKERBSA-N
SMILES
CC1=CC[C@H]2C[C@@H]1C2(C)C
Appearance (Form)
Clear liquid
Refractive index
1.4640 to 1.4680 (20°C, 589 nm)
Specific gravity
(20°C) 0.858 to 0.861
Color scale
=<30 APHA
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text