Thermo Scientific Chemicals

DL-Pipecolinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 131281000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

DL-Pipecolinic acid, 99%, Thermo Scientific Chemicals

Catalog number: 131281000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 131281000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
535-75-1
IUPAC Name
piperidine-2-carboxylic acid
Molecular Formula
C6H11NO2
InChI Key
HXEACLLIILLPRG-UHFFFAOYNA-N
SMILES
OC(=O)C1CCCCN1
Appearance (Form)
Crystalline powder
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Loss on drying
=<1 % (100°C, 4 hrs)
Sulfated ash
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text