Thermo Scientific Chemicals

Piperazine, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 131290100
10 kg, Each
Thermo Scientific Chemicals

Piperazine, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 131290100
10 kg, Each
Quantity
Catalog number: 131290100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
110-85-0
Appearance (Color)
White to light yellow
Infrared spectrum
Conforms
Appearance (Form)
Crystalline flakes
Color of solution
=<30 APHA (50% in methanol)
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text