Piperazine hexahydrate, 98%, Thermo Scientific Chemicals
Piperazine hexahydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Piperazine hexahydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
100 g
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number 131300051
Price (USD)/ Each
438.00
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
438.00
Add to cart
Piperazine hexahydrate, 98%, Thermo Scientific Chemicals
Catalog number131300051
Price (USD)/ Each
438.00
-
Add to cart
Chemical Identifiers
CAS142-63-2
IUPAC Namepiperazine hexahydrate
Molecular FormulaC4H22N2O6
InChI KeyAVRVZRUEXIEGMP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.C1CNCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystals
Infrared spectrumConforms
Titration with HCl>=97.5 %
Appearance (Color)Transparent to white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only