1,3-Propanediol, 98%, Thermo Scientific Chemicals
1,3-Propanediol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Propanediol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 131400010
Price (USD)/ Each
232.65
Online exclusive
274.00 
Save 41.35 (15%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
232.65
Online exclusive
274.00 
Save 41.35 (15%)
Add to cart
1,3-Propanediol, 98%, Thermo Scientific Chemicals
Catalog number131400010
Price (USD)/ Each
232.65
Online exclusive
274.00 
Save 41.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namepropane-1,3-diol
Molecular FormulaC3H8O2
InChI KeyYPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILESOCCCO
Molecular Weight (g/mol)76.10
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<20 APHA
GC>=97.5 %
Infrared spectrumConforms
Water=<0.2 % (K.F.)
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only